4'-Carboxybenzo-15-krone-5 (4'-Carboxybenzo-1,4,7,10,13-pentaoxacyclopentadecan), Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4'-Carboxybenzo-15-crown 5-Ether, 2,3,5,6,8,9,11,12-Octahydrobenzo[b][1,4,7,10,13]pentaoxacyclopentadecine-15-carboxylic acid, (Benzo-15-crown 5-Ether)-4'-carboxylic acid, 2,3-(4-Carboxybenzo)-1,4,7,10,13-pentaoxacyclopentadec-2-ene

21874-1G
SIAL21874-1GEA 286 EUR
SIAL21874-1G
4'-Carboxybenzo-15-krone-5 (4'-Carboxybenzo-1,4,7,10,13-pentaoxacyclopentadecan), Sigma-Aldrich®
4'-Carboxybenzo-15-krone-5 (4'-Carboxybenzo-1,4,7,10,13-pentaoxacyclopentadecan)
Formel: C₁₅H₂₀O₇
Molecular Weight: 312.3151 g/mol
MDL: MFCD00143276
CAS-Nummer: 56683-55-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR