2-Amino-5-methylbenzolsulfonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Aminotoluene-3-sulphonic acid

M-Clarity™ Qualitätsstufe = MQ100

218766-250G
SIAL218766-250GEA 79.1 EUR
SIAL218766-250G
2-Amino-5-methylbenzolsulfonsäure, Sigma-Aldrich®
2-Amino-5-methylbenzenesulphonic acid
Formel: C₇H₉NO₃S
Molecular Weight: 187,22 g/mol
MDL: MFCD00007908
CAS-Nummer: 88-44-8
EINECS: 201-831-3
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR