5(6)-Carboxyfluorescein N-hydroxysuccinimide ester (5/6-FAM SE) ≥80% (durch HPLC), BioReagent geeignet für Fluoreszenz, Sigma-Aldrich®

Lieferant: Merck

21878-100MG-F 21878-25MG-F
SIAL21878-100MG-FEA 164 EUR
SIAL21878-100MG-F SIAL21878-25MG-F
5(6)-Carboxyfluorescein N-hydroxysuccinimide ester (5/6-FAM SE) ≥80% (durch HPLC), BioReagent geeignet für Fluoreszenz, Sigma-Aldrich®
5(6)-Carboxyfluorescein N-hydroxysuccinimide ester (5/6-FAM SE)

5(6)-Carboxyfluorescein N-hydroxysuccinimide ester (5(6)-FAM SE, FLUOS) may be used to prepare fluorescent derivatives of proteins such as tubulin and for the preparation of a phenytoin tracer for the determination of this and other drugs in serum and plasma by fluorescence polarization immunoassay (FPIA) and laser-induced fluorescence detection.


  • Fluorescence λex 492 nm; λem 517 nm in DMF

Formel: C₂₅H₁₅NO₉
MDL: MFCD04119713
CAS-Nummer: 117548-22-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR