Tetraphenylphosphoniumbromid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

218782-100G 218782-25G
SIAL218782-100GEA 236 EUR
SIAL218782-100G SIAL218782-25G
Tetraphenylphosphoniumbromid, Sigma-Aldrich®
Tetraphenylphosphonium bromide
Formel: C₂₄H₂₀BrP
Schmelzpunkt: 299…300 °C
MDL: MFCD00011915
CAS-Nummer: 2751-90-8
EINECS: 220-393-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR