4-(Dimethylamino)zimtsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(Dimethylamino)cinnamic Acid

M-Clarity™ Qualitätsstufe = MQ100

218979-1G
SIAL218979-1GEA 35.8 EUR
SIAL218979-1G
4-(Dimethylamino)zimtsäure, Sigma-Aldrich®
4-(Dimethylamino)zimtsäure
Formel: C₁₁H₁₃NO₂
MDL: MFCD00004397
CAS-Nummer: 1552-96-1
EINECS: 216-299-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR