Carboxymethylcellulose Natriumsalz, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Carboxymethyl cellulose, sodium salt, Cellulose gum, Cellulose, carboxymethyl ether, sodium salt, Sodium carboxymethyl cellulose (Viscosity 800mpa.s), Sodium carboxymethyl cellulose, CMC

21904-1KG
SIAL21904-1KGEA 176 EUR
SIAL21904-1KG
Carboxymethylcellulose Natriumsalz, Sigma-Aldrich®
Carboxymethyl cellulose sodium salt
Formel: [C₆H₇O₂(OH)x(OCH₂COONa)y]n
Lagertemperatur: Raumtemperatur
MDL: MFCD00081472
CAS-Nummer: 9004-32-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR