5(6)-Carboxytetramethylrhodamine succinimidyl ester [5(6)-TAMRA SE], BioReagent geeignet für Fluoreszenz, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (5(6)-TAMRA SE

21955-25MG
SIAL21955-25MGEA 400 EUR
SIAL21955-25MG
5(6)-Carboxytetramethylrhodamine succinimidyl ester [5(6)-TAMRA SE], BioReagent geeignet für Fluoreszenz, Sigma-Aldrich®
5(6)-Carboxytetramethylrhodamine succinimidyl ester [5(6)-TAMRA SE]

5(6)-Carboxytetramethylrhodamine N-succinimidyl ester (TMR-SE) is used as an amine coupling reagent to form 5(6)-carboxytetramethylrhodamine (TMR) derivatized compounds such as proteins and drugs.


  • Assay ≥70% (coupling to amines)
  • Fluorescence λex 543 nm; λem 576 nm in methanol


Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.

Formel: C₂₉H₂₅N₃O₇
Molecular Weight: 527,52 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00077324
CAS-Nummer: 150408-83-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR