(±)-2-Hydroxyisovaleriansäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Hydroxy-3-methylbutanoic acid, 2-Hydroxy-3-methylbutyric acid

M-Clarity™ Qualitätsstufe = MQ200

219835-1G
SIAL219835-1GEA 190 EUR
SIAL219835-1G
(±)-2-Hydroxyisovaleriansäure, Sigma-Aldrich®
(±)-2-Hydroxyisovaleriansäure
Formel: C₅H₁₀O₃
Molecular Weight: 118,13 g/mol
MDL: MFCD00004242
CAS-Nummer: 4026-18-0
EINECS: 223-697-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR