3-Hydroxybenzophenon, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
m-Hydroxybenzophenon
220434-1G
SIAL220434-1GEA
82.4
EUR
SIAL220434-1G
3-Hydroxybenzophenon, Sigma-Aldrich®
3-Hydroxybenzophenon
Formel:
C₁₃H₁₀O₂ Molecular Weight: 198,22 g/mol Siedepunkt: 148 °C (4 mmHg) Schmelzpunkt: 117 °C |
MDL:
MFCD00002297 CAS-Nummer: 13020-57-0 EINECS: 235-879-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...