Thiophen-2-methylamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Thiophenemethylamine, 2-(Aminomethyl)thiophene

M-Clarity™ Qualitätsstufe = MQ200

220884-5G
SIAL220884-5GEA 72.7 EUR
SIAL220884-5G
Thiophen-2-methylamin, Sigma-Aldrich®
Thiophen-2-methylamin
Formel: C₅H₇NS
Molecular Weight: 113.19 g/mol
MDL: MFCD00005460
CAS-Nummer: 27757-85-3
EINECS: 248-639-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR