1,4-Butandioldiglycidylether, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Araldite RD-2
M-Clarity™ Qualitätsstufe = MQ200
220892-10G
220892-250G
220892-50G
SIAL220892-10GEA
103
EUR
SIAL220892-10G
SIAL220892-250G
SIAL220892-50G
1,4-Butandioldiglycidylether, Sigma-Aldrich®
1,4-Butandioldiglycidylether
Formel:
C₁₀H₁₈O₄ Molecular Weight: 202.25 g/mol |
MDL:
MFCD00005146 CAS-Nummer: 2425-79-8 EINECS: 219-371-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...