4'-Bromvalerophenon, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

221414-10G
SIAL221414-10GEA 81 EUR
SIAL221414-10G
4'-Bromvalerophenon, Sigma-Aldrich®
4'-Bromvalerophenon
Formel: C₁₁H₁₃BrO
Molecular Weight: 241,13 g/mol
MDL: MFCD00000107
CAS-Nummer: 7295-44-5
EINECS: 230-729-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR