Bis(triphenylphosphin)kupfer(I)borhydrid, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Copper bis(triphenylphosphine)(tetrahydroborate), Copper bis(triphenylphosphine)(borhydride), Tris(triphenylphosphine)copper (I) tetrahydroborate

M-Clarity™ Qualitätsstufe = MQ100

222364-5G
SIAL222364-5GEA 78.1 EUR
SIAL222364-5G
Bis(triphenylphosphin)kupfer(I)borhydrid, Sigma-Aldrich®
Bis(triphenylphosphin)kupfer(I)borhydrid
Formel: C₃₆H₃₄BCuP₂
Molecular Weight: 602,97 g/mol
MDL: MFCD00013219
CAS-Nummer: 16903-61-0
EINECS: 240-951-0
UN: 2813
ADR: 4.3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR