5-Methylisatin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Methyl-2,3-dioxoindolin, 5-Methyl-1H-indol-2,3-dion, 5-Methylindolin-2,3-dion, 5-Methyl-2,3-indolindion

M-Clarity™ Qualitätsstufe = MQ100

222429-5G
SIAL222429-5GEA 105 EUR
SIAL222429-5G
5-Methylisatin, Sigma-Aldrich®
5-Methylisatin
Formel: C₉H₇NO₂
Molecular Weight: 161,16 g/mol
MDL: MFCD00005721
CAS-Nummer: 608-05-9
EINECS: 210-152-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR