Bariumiodid Hydrat, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Barium iodide dihydrate
M-Clarity™ Qualitätsstufe = MQ100
223808-50G
SIAL223808-50GEA
192
EUR
SIAL223808-50G
Bariumiodid Hydrat, Sigma-Aldrich®
Bariumiodid Hydrat
Formel:
BaI₂·H₂O Molecular Weight: 409,15 g/mol Dichte: 5,15 g/cm³ (20 °C) |
MDL:
MFCD00149155 CAS-Nummer: 7787-33-9 EINECS: 237-276-9 UN: 1564 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...