(1,3-Dioxolan-2-ylmethyl)triphenylphosphoniumbromid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: ((1,3-Dioxolan-2-yl)methyl)triphenylphosphonium bromide

M-Clarity™ Qualitätsstufe = MQ200

223859-5G
SIAL223859-5GEA 34.4 EUR
SIAL223859-5G
(1,3-Dioxolan-2-ylmethyl)triphenylphosphoniumbromid, Sigma-Aldrich®
(1,3-Dioxolan-2-ylmethyl)triphenylphosphoniumbromid
Formel: C₂₂H₂₂BrO₂P
Molecular Weight: 429,30 g/mol
MDL: MFCD00011966
CAS-Nummer: 52509-14-5
EINECS: 257-977-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR