(3-Fluor-4-hydroxyphenyl)essigsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Fluoro-4-hydroxyphenylacetic acid

M-Clarity™ Qualitätsstufe = MQ100

224510-1G
SIAL224510-1GEA 71.1 EUR
SIAL224510-1G
(3-Fluor-4-hydroxyphenyl)essigsäure, Sigma-Aldrich®
(3-Fluor-4-hydroxyphenyl)essigsäure
Formel: C₈H₇FO₃
Molecular Weight: 170,14 g/mol
MDL: MFCD00004348
CAS-Nummer: 458-09-3
EINECS: 207-275-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR