2-(Phenylsulfonyl)anilin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

225045-1G
SIAL225045-1GEA 49.2 EUR
SIAL225045-1G
2-(Phenylsulfonyl)anilin, Sigma-Aldrich®
2-(Phenylsulfonyl)anilin
Formel: C₁₂H₁₁NO₂S
Molecular Weight: 233,2862 g/mol
MDL: MFCD00007706
CAS-Nummer: 4273-98-7
EINECS: 224-271-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR