2-Brombenzylamin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

225169-5G
SIAL225169-5GEA 80.4 EUR
SIAL225169-5G
2-Brombenzylamin Hydrochlorid, Sigma-Aldrich®
2-Brombenzylamin Hydrochlorid
Formel: C₇H₉BrClN
Molecular Weight: 222,52 g/mol
MDL: MFCD00012853
CAS-Nummer: 5465-63-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR