2-Chlor-4,6-dinitroanilin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
6-Chlor-2,4-dinitroanilin
M-Clarity™ Qualitätsstufe = MQ100
225428-100G
SIAL225428-100GEA
52.6
EUR
SIAL225428-100G
2-Chlor-4,6-dinitroanilin, Sigma-Aldrich®
2-Chlor-4,6-dinitroanilin
Formel:
C₆H₄ClN₃O₄ Molecular Weight: 217,57 g/mol Schmelzpunkt: 248 °C |
MDL:
MFCD00007147 CAS-Nummer: 3531-19-9 EINECS: 222-564-9 UN: 2237 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...