3-Acetyl-2,5-dimethylthiophen, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

225797-10G
SIAL225797-10GEA 56.8 EUR
SIAL225797-10G
3-Acetyl-2,5-dimethylthiophen, Sigma-Aldrich®
3-Acetyl-2,5-dimethylthiophen
Formel: C₈H₁₀OS
Molecular Weight: 154,23 g/mol
MDL: MFCD00009763
CAS-Nummer: 2530-10-1
EINECS: 219-779-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR