1,2-Naphthochinon-4-sulfonsäure Natriumsalz, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Folin's reagent, Sodium 3,4-dioxo-3,4-dihydronaphthalene-1-sulphonate

M-Clarity™ Qualitätsstufe = MQ100

226017-10G
SIAL226017-10GEA 159 EUR
SIAL226017-10G
1,2-Naphthochinon-4-sulfonsäure Natriumsalz, Sigma-Aldrich®
1,2-Naphthochinon-4-sulfonsäure Natriumsalz
Formel: C₁₀H₅NaO₅S
Molecular Weight: 260,20 g/mol
MDL: MFCD00001700
CAS-Nummer: 521-24-4
EINECS: 208-308-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR