tert-Butyl-N-hydroxycarbamat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-Boc-hydroxylamine, N-tert-Butoxycarbonylhydroxylamine, N-Tert-Boc-Hydroxylamine

M-Clarity™ Qualitätsstufe = MQ200

226157-5G
SIAL226157-5GEA 91.9 EUR
SIAL226157-5G
tert-Butyl-N-hydroxycarbamat, Sigma-Aldrich®
tert-Butyl-N-hydroxycarbamat
Formel: C₅H₁₁NO₃
Molecular Weight: 133,15 g/mol
MDL: MFCD00002107
CAS-Nummer: 36016-38-3
EINECS: 252-836-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR