(1S)-(-)-Camphansäurechlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(1S)-4,7,7-Trimethyl-3-oxo-2-oxabicyclo[2.2.1]heptane-1-carbonyl chloride, (-)-Camphanic acid chloride
M-Clarity™ Qualitätsstufe = MQ200
226173-1G
226173-5G
SIAL226173-1GEA
45.9
EUR
SIAL226173-1G
SIAL226173-5G
(1S)-(-)-Camphansäurechlorid, Sigma-Aldrich®
(1S)-(-)-Camphansäurechlorid
Formel:
C₁₀H₁₃ClO₃ Molecular Weight: 216.67 g/mol |
MDL:
MFCD00135626 CAS-Nummer: 39637-74-6 EINECS: 254-552-4 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...