(-)-Chinin Hydrochlorid Dihydrat, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: Quinine hydrochloride dihydrate, Quinine monohydrochloride dihydrate, (R)-(6-Methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methanol hydrochloride dihydrate

M-Clarity™ Qualitätsstufe = MQ100

22632-50G-F
SIAL22632-50G-FEA 211 EUR
SIAL22632-50G-F
(-)-Chinin Hydrochlorid Dihydrat, Sigma-Aldrich®
(-)-Chinin Hydrochlorid Dihydrat
Formel: C₂₀H₂₅ClN₂O₂·2H₂O
Molecular Weight: 396,91 g/mol
Schmelzpunkt: 158…160 °C
Flammpunkt: 122 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00151248
CAS-Nummer: 6119-47-7
EINECS: 205-001-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR