Quinine hemisulphate monohydrate Ph. Eur., Sigma-Aldrich®

Lieferant: Merck

Synonyme: Quinine sulphate, Quinine sulphate (2:1) (salt) dihydrate

M-Clarity™ Qualitätsstufe = MQ100

22642-50G
SIAL22642-50GEA 226 EUR
SIAL22642-50G
Quinine hemisulphate monohydrate Ph. Eur., Sigma-Aldrich®
Quinine hemisulphate monohydrate
Formel: C₂₀H₂₄N₂O₂·0.5H₂SO₄·H₂O
Molecular Weight: 391,47 g/mol (373,46 anhy)
Schmelzpunkt: 219 °C
Dichte: 1,36 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00150792
CAS-Nummer: 6119-70-6
EINECS: 212-359-2
Merck Index: 12,03822

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR