2-(Methylthio)ethanol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

226424-10G
SIAL226424-10GEA 40.5 EUR
SIAL226424-10G
2-(Methylthio)ethanol, Sigma-Aldrich®
2-(Methylthio)ethanol
Formel: C₃H₈OS
Molecular Weight: 92.16 g/mol
MDL: MFCD00002908
CAS-Nummer: 5271-38-5
EINECS: 226-090-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR