(Phenylthio)trimethylsilan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

226467-5G
SIAL226467-5GEA 124 EUR
SIAL226467-5G
(Phenylthio)trimethylsilan, Sigma-Aldrich®
(Phenylthio)trimethylsilane
Formel: C₉H₁₄SSi
MDL: MFCD00008272
CAS-Nummer: 4551-15-9
EINECS: 224-916-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR