1,1,3,3-Tetramethylbutylisocyanid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,1,3,3-Tetramethylbutylisocyanide

M-Clarity™ Qualitätsstufe = MQ200

226491-5G
SIAL226491-5GEA 177 EUR
SIAL226491-5G
1,1,3,3-Tetramethylbutylisocyanid, Sigma-Aldrich®
1,1,3,3-Tetramethylbutylisocyanid
Formel: C₉H₁₇N
Molecular Weight: 139,2380 g/mol
MDL: MFCD00000003
CAS-Nummer: 14542-93-9
EINECS: 238-577-8
UN: 3275
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR