Silberperchlorat Hydrat ≥99%, Sigma-Aldrich®
Lieferant: Merck
226548-25G
SIAL226548-25GEA
207
EUR
SIAL226548-25G
Silberperchlorat Hydrat ≥99%, Sigma-Aldrich®
Silberperchlorat Hydrat
Formel:
AgClO₄·xH₂O |
MDL:
MFCD00149128 CAS-Nummer: 331717-44-3 EINECS: 232-035-4 UN: 1481 ADR: 5.1,II Merck Index: 12,08667 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...