(±)-1-Phenyl-2-propin-1-ol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Phenyl-2-propyn-1-ol

M-Clarity™ Qualitätsstufe = MQ100

226610-10G 226610-1G
SIAL226610-10GEA 190 EUR
SIAL226610-10G SIAL226610-1G
(±)-1-Phenyl-2-propin-1-ol, Sigma-Aldrich®
(±)-1-Phenyl-2-propin-1-ol
Formel: C₉H₈O
Molecular Weight: 132.16 g/mol
MDL: MFCD00021860
CAS-Nummer: 4187-87-5
EINECS: 224-064-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR