(±)-1-Phenyl-2-propin-1-ol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-Phenyl-2-propyn-1-ol
M-Clarity™ Qualitätsstufe = MQ100
226610-10G
226610-1G
SIAL226610-10GEA
190
EUR
SIAL226610-10G
SIAL226610-1G
(±)-1-Phenyl-2-propin-1-ol, Sigma-Aldrich®
(±)-1-Phenyl-2-propin-1-ol
Formel:
C₉H₈O Molecular Weight: 132.16 g/mol |
MDL:
MFCD00021860 CAS-Nummer: 4187-87-5 EINECS: 224-064-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...