1-(Trimethylsiloxy)-1,3-butadien, Sigma-Aldrich®

Lieferant: Merck

227226-10G 227226-50G
SIAL227226-10GEA 138 EUR
SIAL227226-10G SIAL227226-50G
1-(Trimethylsiloxy)-1,3-butadien, Sigma-Aldrich®
1-(Trimethylsiloxy)-1,3-butadien
Formel: C₇H₁₄OSi
CAS-Nummer: 6651-43-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR