1-Diazo-2-naphthol-4-sulfonsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

227285-100G
SIAL227285-100GEA 83.4 EUR
SIAL227285-100G
1-Diazo-2-naphthol-4-sulfonsäure, Sigma-Aldrich®
1-Diazo-2-naphthol-4-sulfonsäure
Formel: C₁₀H₆N₂O₄S
Molecular Weight: 250.23 g/mol
MDL: MFCD00012409
CAS-Nummer: 887-76-3
EINECS: 212-958-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR