4-Brombutylacetat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Essigsäure(4-brombutyl)ester

M-Clarity™ Qualitätsstufe = MQ100

227579-10G 227579-50G
SIAL227579-10GEA 57.6 EUR
SIAL227579-10G SIAL227579-50G
4-Brombutylacetat, Sigma-Aldrich®
4-Brombutylacetat
Formel: C₆H₁₁BrO₂
MDL: MFCD00000263
CAS-Nummer: 4753-59-7
EINECS: 225-277-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR