Phenylhydrochinon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: TFIIH Modulator-12, Biphenyl-2,5-diol, [1,1'-Biphenyl]-2,5-diol, 2,5-Dihydroxybiphenyl

M-Clarity™ Qualitätsstufe = MQ100

227811-25G
SIAL227811-25GEA 113 EUR
SIAL227811-25G
Phenylhydrochinon, Sigma-Aldrich®
Phenylhydrochinon
Formel: C₁₂H₁₀O₂
Molecular Weight: 186,21 g/mol
MDL: MFCD00002342
CAS-Nummer: 1079-21-6
EINECS: 214-091-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR