(Dimethylamino)acetonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

228540-25G
SIAL228540-25GEA 0 EUR
SIAL228540-25G
(Dimethylamino)acetonitril, Sigma-Aldrich®
(Dimethylamino)acetonitril
Formel: C₄H₈N₂
Molecular Weight: 84,12 g/mol
MDL: MFCD00001890
CAS-Nummer: 926-64-7
EINECS: 213-140-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR