2-(2-Fluorphenyl)ethanol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Fluorophenethyl alcohol
M-Clarity™ Qualitätsstufe = MQ100
228788-5G
SIAL228788-5GEA
47.5
EUR
SIAL228788-5G
2-(2-Fluorphenyl)ethanol, Sigma-Aldrich®
2-(2-Fluorphenyl)ethanol
Formel:
C₈H₉FO Molecular Weight: 140,16 g/mol Siedepunkt: 104…106 °C (14 mmHg) Dichte: 1,04 g/cm³ (20 °C) |
MDL:
MFCD00002887 CAS-Nummer: 50919-06-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...