Diethylpropylmalonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Diethyl 2-propylmalonate

M-Clarity™ Qualitätsstufe = MQ200

228818-5G
SIAL228818-5GEA 28.1 EUR
SIAL228818-5G
Diethylpropylmalonat, Sigma-Aldrich®
Diethylpropylmalonat
Formel: C₁₀H₁₈O₄
Molecular Weight: 202,25 g/mol
MDL: MFCD00009168
CAS-Nummer: 2163-48-6
EINECS: 218-492-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR