Platin(IV)oxid Monohydrat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
229040-1G
SIAL229040-1GEA
208
EUR
SIAL229040-1G
Platin(IV)oxid Monohydrat, Sigma-Aldrich®
Platin(IV)oxid Monohydrat
Formel:
O₂Pt·1H₂O Molecular Weight: 245.09 g/mol |
MDL:
MFCD00066964 CAS-Nummer: 12137-21-2 EINECS: 215-223-0 UN: 1479 ADR: 5.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...