Cobalt(II)sulfat Hydrat, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
Synonyme:
Cobaltsulfat Hydrat
M-Clarity™ Qualitätsstufe = MQ100
229598-10G
SIAL229598-10GEA
186
EUR
SIAL229598-10G
Cobalt(II)sulfat Hydrat, Sigma-Aldrich®
Cobalt(II)sulfat Hydrat
Formel:
CoO₄S·H₂O |
MDL:
MFCD00149657 CAS-Nummer: 60459-08-7 EINECS: 233-334-2 UN: 3077 ADR: 9,III Merck Index: 12,02510 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...