2,4-Diaminophenol Dihydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Hydroxy-1,3-Phenylenediammonium Dichloride, 2,4-Diaminophenol Dihydrochloride, Amidol

M-Clarity™ Qualitätsstufe = MQ100

230103-100G
SIAL230103-100GEA 479 EUR
SIAL230103-100G
2,4-Diaminophenol Dihydrochlorid, Sigma-Aldrich®
2,4-Diaminophenol Dihydrochlorid
Formel: C₆H₁₀Cl₂N₂O
Molecular Weight: 197.06 g/mol
MDL: MFCD00012979
CAS-Nummer: 137-09-7
EINECS: 205-279-4
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR