Kupfer(I)bromid-Dimethylsulfid-Komplex, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
230502-10G
230502-50G
SIAL230502-10GEA
100
EUR
SIAL230502-10G
SIAL230502-50G
Kupfer(I)bromid-Dimethylsulfid-Komplex, Sigma-Aldrich®
Kupfer(I)bromid-Dimethylsulfid-Komplex
Formel:
C₂H₆BrCuS |
MDL:
MFCD00043295 CAS-Nummer: 54678-23-8 EINECS: 259-292-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...