4-(Methylthio)benzylalkohol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(Methylthio)benzyl Alcohol

230685-10G
SIAL230685-10GEA 129 EUR
SIAL230685-10G
4-(Methylthio)benzylalkohol, Sigma-Aldrich®
4-(Methylthio)benzylalkohol
Formel: C₈H₁₀OS
Molecular Weight: 154,23 g/mol
MDL: MFCD00009706
CAS-Nummer: 3446-90-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR