1,3-Diphenoxybenzol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

232637-5G
SIAL232637-5GEA 59.5 EUR
SIAL232637-5G
1,3-Diphenoxybenzol, Sigma-Aldrich®
1,3-Diphenoxybenzene
Formel: C₁₈H₁₄O₂
Molecular Weight: 262,31 g/mol
MDL: MFCD00003036
CAS-Nummer: 3379-38-2
EINECS: 222-181-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR