2-Hydroxyethyl-phenylsulfid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(Phenylthio)ethanol

M-Clarity™ Qualitätsstufe = MQ100

232777-50G
SIAL232777-50GEA 95.7 EUR
SIAL232777-50G
2-Hydroxyethyl-phenylsulfid, Sigma-Aldrich®
2-Hydroxyethyl-phenylsulfid
Formel: C₈H₁₀OS
Molecular Weight: 154,23 g/mol
MDL: MFCD00002905
CAS-Nummer: 699-12-7
EINECS: 211-828-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR