2-(3-Chlorphenoxy)propionsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

233013-50G
SIAL233013-50GEA 71.1 EUR
SIAL233013-50G
2-(3-Chlorphenoxy)propionsäure, Sigma-Aldrich®
2-(3-Chlorphenoxy)propionsäure
Formel: C₉H₉ClO₃
MDL: MFCD00009723
CAS-Nummer: 101-10-0
EINECS: 202-915-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR