(α,α,α-Trifluor-4-tolyl)essigsäure, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: (4-(Trifluormethyl)phenyl)essigsäure

M-Clarity™ Qualitätsstufe = MQ200

233021-5G
SIAL233021-5GEA 58.6 EUR
SIAL233021-5G
(α,α,α-Trifluor-4-tolyl)essigsäure, Sigma-Aldrich®
(α,α,α-Trifluor-4-tolyl)essigsäure
Formel: (C₆H₄(CF₃))CH₂COOH
Molecular Weight: 204,15 g/mol
MDL: MFCD00004352
CAS-Nummer: 32857-62-8
EINECS: 251-263-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR