trans-4-(Trifluormethyl)zimtsäure, Sigma-Aldrich®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ200

233099-5G
SIAL233099-5GEA 95.8 EUR
SIAL233099-5G
trans-4-(Trifluormethyl)zimtsäure, Sigma-Aldrich®
trans-4-(Trifluormethyl)zimtsäure
Formel: C₁₀H₇F₃O₂
Molecular Weight: 216,16 g/mol
MDL: MFCD00002696
CAS-Nummer: 16642-92-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR