2-Chlor-1-fluor-4-nitrobenzol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3-Chlor-4-fluornitrobenzol, 1-Chlor-2-fluor-5-nitrobenzol
233234-25G
SIAL233234-25GEA
148
EUR
SIAL233234-25G
2-Chlor-1-fluor-4-nitrobenzol, Sigma-Aldrich®
2-Chlor-1-fluor-4-nitrobenzol
Formel:
C₆H₃ClFNO₂ Molecular Weight: 175,55 g/mol Siedepunkt: 227…228 °C (1013 hPa) Schmelzpunkt: 41,5 °C |
MDL:
MFCD00007206 CAS-Nummer: 350-30-1 EINECS: 206-499-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...